accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 584-42-9

IUPAC Name: sodium 3-[2-(3-nitrophenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
Synonyms: sodium 3-[2-(3-nitrophenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
Molecular Weight (g/mol): 309.21
Molecular Formula: C13H8N3NaO5
InChi Key: GCGHYYRSTZNXJS-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC(C=CC1=O)=NNC1=CC(=CC=C1)[N+]([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results