accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHSOHOCH3OO(E)(E)

CAS RN 58416-04-9

IUPAC Name: (2E,5E)-bis[hydroxy(methoxy)methylidene]thiolane-3,4-dione
Synonyms: (2E,5E)-bis[hydroxy(methoxy)methylidene]thiolane-3,4-dione
Molecular Weight (g/mol): 232.21
Molecular Formula: C8H8O6S
InChi Key: DGAOBLHPQCJULL-KQQUZDAGSA-N
SMILES: CO\C(O)=C1\S\C(=C(/O)OC)C(=O)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results