accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5845-53-4

IUPAC Name: methyl (2R)-2-amino-4-methylpentanoate hydrochloride
Synonyms: —
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: DODCBMODXGJOKD-HQFIOVGONA-N
SMILES: Cl.COC(=O)[C@H](N)CC(C)C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)