accessibility menu, dialog, popup

UserName

Viewing profile as user:
POOPO

CAS RN 5849-36-5

IUPAC Name: diphenylphosphoroso diphenylphosphinate
Synonyms: diphenylphosphoroso diphenylphosphinate
Molecular Weight (g/mol): 418.37
Molecular Formula: C24H20O3P2
InChi Key: XTAYANNRWHXQOL-UHFFFAOYSA-N
SMILES: O=P(OP(=O)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results