CAS RN 58491-62-6
IUPAC Name:
furan-2,5-dicarbonitrile
Synonyms:
furan-2,5-dicarbonitrile
Molecular Weight (g/mol):
118.1
Molecular Formula:
C6H2N2O
InChi Key:
KCSYJHQYWTYFCM-UHFFFAOYSA-N
SMILES:
N#CC1=CC=C(O1)C#N
1
–
1
of
1
results
1
2,5-Dicyanofuran 98.0+%, TCI America™
CAS: 58491-62-6 Molecular Formula: C6H2N2O Molecular Weight (g/mol): 118.095 MDL Number: MFCD00671519 InChI Key: KCSYJHQYWTYFCM-UHFFFAOYSA-N Synonym: 2,5-Furandicarbonitrile PubChem CID: 94081 IUPAC Name: furan-2,5-dicarbonitrile SMILES: C1=C(OC(=C1)C#N)C#N