accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 585-28-4

IUPAC Name: butyl carbamoylformate
Synonyms: butyl carbamoylformate
Molecular Weight (g/mol): 145.16
Molecular Formula: C6H11NO3
InChi Key: JQOLMUGUXNNIOR-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results