accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 585-29-5

IUPAC Name: formic acid; triethylamine
Synonyms: formic acid; triethylamine
Molecular Weight (g/mol): 147.22
Molecular Formula: C7H17NO2
InChi Key: PTMFUWGXPRYYMC-UHFFFAOYSA-N
SMILES: OC=O.CCN(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results