accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 585-74-0

IUPAC Name: 1-(3-methylphenyl)ethan-1-one
Synonyms: M-methylacetophenone
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: FSPSELPMWGWDRY-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(C)=C1
Boiling Point 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)