accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 585-79-5

CAS RN 585-79-5

IUPAC Name: 1-bromo-3-nitrobenzene
Synonyms: 3-bromo-1-nitrobenzene
Molecular Weight (g/mol): 202.01
Molecular Formula: C6H4BrNO2
InChi Key: FWIROFMBWVMWLB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(Br)=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results