accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOOHNNONHNSOOOO(E)(E)

CAS RN 5850-16-8

IUPAC Name: disodium 4-{2-[(1E,5E)-4,6-dioxo-5-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]cyclohex-2-en-1-ylidene]hydrazin-1-yl}naphthalene-1-sulfonate
Synonyms: disodium 4-{2-[(1E,5E)-4,6-dioxo-5-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]cyclohex-2-en-1-ylidene]hydrazin-1-yl}naphthalene-1-sulfonate
Molecular Weight (g/mol): 622.53
Molecular Formula: C26H16N4Na2O8S2
InChi Key: UTIACSXEVKXHPT-XALLQCHJSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=C2C=CC=CC2=C(N\N=C2/C=CC(=O)\C(=N/NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results