accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5854-77-3

OOONH2(R)(S)CH3CH3CH3H3CH3CH3CCH3OHOH3C

CAS RN 5854-77-3

IUPAC Name: acetic acid; tert-butyl (2S,3R)-2-amino-3-(tert-butoxy)butanoate
Synonyms: acetic acid; tert-butyl (2S,3R)-2-amino-3-(tert-butoxy)butanoate
Molecular Weight (g/mol): 291.39
Molecular Formula: C14H29NO5
InChi Key: BGAUVMFJRASONL-RJUBDTSPSA-N
SMILES: CC(O)=O.C[C@@H](OC(C)(C)C)[C@H](N)C(=O)OC(C)(C)C
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)