accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58546-89-7

IUPAC Name: 1-benzofuran-5-amine
Synonyms: 1-benzofuran-5-amine
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: GMOLCSICTCPZCU-UHFFFAOYSA-N
SMILES: NC1=CC=C2OC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results