accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58551-83-0

IUPAC Name: 2,4,6-trifluorobenzaldehyde
Synonyms: 2,4,6-trifluorobenzaldehyde
Molecular Weight (g/mol): 160.1
Molecular Formula: C7H3F3O
InChi Key: KPJIEPBITZLHPQ-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C=O)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results