CAS RN 58561-04-9
IUPAC Name:
methyl 2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoate
Synonyms:
methyl 2-[(tert-butoxycarbonyl)amino]-3-methylbutanoate
Molecular Weight (g/mol):
231.29
Molecular Formula:
C11H21NO4
InChi Key:
XCJLIYKAMLUDGN-UHFFFAOYNA-N
SMILES:
COC(=O)C(NC(=O)OC(C)(C)C)C(C)C
1
–
1
of
1
results
1
Sigma Aldrich Ruthenium(Iv) Oxide Hydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.