accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBr

CAS RN 58573-93-6

IUPAC Name: 4-bromo-4'-heptyl-1,1'-biphenyl
Synonyms: 4-bromo-4'-heptyl-1,1'-biphenyl
Molecular Weight (g/mol): 331.3
Molecular Formula: C19H23Br
InChi Key: RJQRJLCQHIMUQO-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results