accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH

CAS RN 58573-94-7

IUPAC Name: 4'-heptyl-[1,1'-biphenyl]-4-carboxylic acid
Synonyms: 4'-heptyl-[1,1'-biphenyl]-4-carboxylic acid
Molecular Weight (g/mol): 296.41
Molecular Formula: C20H24O2
InChi Key: XVROSBHWACAQRL-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results