accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaCH3NHNOOOSOOO(E)

CAS RN 5858-81-1

IUPAC Name: disodium (4E)-4-[2-(4-methyl-2-sulfonatophenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-2-carboxylate
Synonyms: pigment red 57
Molecular Weight (g/mol): 430.34
Molecular Formula: C18H12N2Na2O6S
InChi Key: CTBASIXHDUUSCV-SDXCCTRSSA-L
SMILES: [Na+].[Na+].CC1=CC=C(N\N=C2\C(=O)C(=CC3=CC=CC=C23)C([O-])=O)C(=C1)S([O-])(=O)=O
Color 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)