accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NClCl

CAS RN 58584-94-4

IUPAC Name: 2,6-dichloro-3-methylpyridine
Synonyms: 2,6-dichloro-3-methylpyridine
Molecular Weight (g/mol): 162.01
Molecular Formula: C6H5Cl2N
InChi Key: KNIKCOPEHZTQJV-UHFFFAOYSA-N
SMILES: CC1=C(Cl)N=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results