CAS RN 5859-93-8
IUPAC Name:
[6-(hydroxymethyl)naphthalen-2-yl]methanol
Synonyms:
[6-(hydroxymethyl)naphthalen-2-yl]methanol
Molecular Weight (g/mol):
188.23
Molecular Formula:
C12H12O2
InChi Key:
SSFGHKDDKYEERH-UHFFFAOYSA-N
SMILES:
OCC1=CC2=CC=C(CO)C=C2C=C1
1
–
1
of
1
results
1
2,6-Bis(hydroxymethyl)naphthalene 98.0+%, TCI America™
CAS: 5859-93-8 Molecular Formula: C12H12O2 Molecular Weight (g/mol): 188.226 InChI Key: SSFGHKDDKYEERH-UHFFFAOYSA-N PubChem CID: 11788722 IUPAC Name: [6-(hydroxymethyl)naphthalen-2-yl]methanol SMILES: C1=CC2=C(C=CC(=C2)CO)C=C1CO