accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3

CAS RN 586-62-9

IUPAC Name: 1-methyl-4-(propan-2-ylidene)cyclohex-1-ene
Synonyms: terpinolene fema number 3046 nofmer TP terpinolen 1,4(8)-terpadiene α-terpinolene tereben isoterpinene 1,4(8)-p-menthadiene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: MOYAFQVGZZPNRA-UHFFFAOYSA-N
SMILES: CC(C)=C1CCC(C)=CC1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)