accessibility menu, dialog, popup

UserName

Viewing profile as user:
ON

CAS RN 586-96-9

IUPAC Name: nitrosobenzene
Synonyms: nitrosobenzene nitrosobenzene, 14C-labeled NOB 14C-labeled nitrosobenzene nitrosobenzene, 14c-labeled 14c-labeled nitrosobenzene
Molecular Weight (g/mol): 107.11
Molecular Formula: C6H5NO
InChi Key: NLRKCXQQSUWLCH-UHFFFAOYSA-N
SMILES: O=NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)