CAS RN 58609-73-7
CAS RN 58609-73-7
IUPAC Name:
2-[4-(2-methylpropyl)phenyl]propanenitrile
Synonyms:
2-[4-(2-methylpropyl)phenyl]propanenitrile
Molecular Weight (g/mol):
187.29
Molecular Formula:
C13H17N
InChi Key:
PKQKHWNHCKNYSW-UHFFFAOYNA-N
SMILES:
CC(C)CC1=CC=C(C=C1)C(C)C#N
1
–
1
of
1
results
1
Thermo Scientific Chemicals 2-(4-Isobutylphenyl)propanenitrile, 97%, Thermo Scientific™
CAS: 58609-73-7 Molecular Formula: C13H17N Molecular Weight (g/mol): 187.286 MDL Number: MFCD02180706 InChI Key: PKQKHWNHCKNYSW-UHFFFAOYSA-N Synonym: 2-4-isobutylphenyl propanenitrile,2-4-isobutylphenyl propionitrile,2-4-2-methylpropyl phenyl propanenitrile,2-4-isobutylphenyl propiononitrile,2-p-isobutylphenyl propiononitrile,+--2-4'-isobutylphenyl propionitrile,+/--2-4'-isobutylphenyl propionitrile,ibuprofen nitrile,aronis24266,2-4-isobutylphenyl-propionitrile PubChem CID: 108374 ChEBI: CHEBI:27415 IUPAC Name: 2-[4-(2-methylpropyl)phenyl]propanenitrile SMILES: CC(C)CC1=CC=C(C=C1)C(C)C#N