accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 586394-64-1

IUPAC Name: N-(4-bromo-3-methylphenyl)-2-methoxybenzamide
Synonyms: N-(4-bromo-3-methylphenyl)-2-methoxybenzamide
Molecular Weight (g/mol): 320.19
Molecular Formula: C15H14BrNO2
InChi Key: DAJGBNVYZLWLSV-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)NC1=CC(C)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results