accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3O

CAS RN 58654-67-4

IUPAC Name: 5-methyloctan-2-one
Synonyms: 2-octanone, 5-methyl-
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: UPVLGOPSYZTLGB-UHFFFAOYNA-N
SMILES: CCCC(C)CCC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results