accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5867-98-1

IUPAC Name: dipentyl sulfate
Synonyms: dipentyl sulfate
Molecular Weight (g/mol): 238.34
Molecular Formula: C10H22O4S
InChi Key: GAFRWLVTHPVQGK-UHFFFAOYSA-N
SMILES: CCCCCOS(=O)(=O)OCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results