accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClO

CAS RN 587-04-2

IUPAC Name: 3-chlorobenzaldehyde
Synonyms: M-chlorobenzaldehyde
Molecular Weight (g/mol): 140.57
Molecular Formula: C7H5ClO
InChi Key: SRWILAKSARHZPR-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(C=O)=C1
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (4)
Percent Purity 
  • (3)
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (6)
  • (4)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)