accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3OHNOO

CAS RN 587-48-4

IUPAC Name: sodium 3-acetamidobenzoate
Synonyms: sodium 3-acetamidobenzoate
Molecular Weight (g/mol): 201.16
Molecular Formula: C9H8NNaO3
InChi Key: FEMJCNCAWASVDS-UHFFFAOYSA-M
SMILES: [Na+].CC(=O)NC1=CC=CC(=C1)C([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)