accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 587-84-8

IUPAC Name: N-phenylaniline; sulfuric acid
Synonyms: diphenylamine; sulfuric acid
Molecular Weight (g/mol): 267.3
Molecular Formula: C12H13NO4S
InChi Key: IPZMDJYHJNHGML-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results