accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5872-08-2

H3CH3CSHOOOO

CAS RN 5872-08-2

IUPAC Name: {7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl}methanesulfonic acid
Synonyms: camphorsulfonic acid reychler's acid CSA camphersulfosaeure
Molecular Weight (g/mol): 232.29
Molecular Formula: C10H16O4S
InChi Key: MIOPJNTWMNEORI-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(CS(O)(=O)=O)C(=O)C2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results