CAS RN 58734-57-9
IUPAC Name:
3-methoxybenzene-1-sulfonamide
Synonyms:
benzenesulfonamide, 3-methoxy-
Molecular Weight (g/mol):
187.21
Molecular Formula:
C7H9NO3S
InChi Key:
VBKIEQKVSHDVGH-UHFFFAOYSA-N
SMILES:
COC1=CC(=CC=C1)S(N)(=O)=O
1
–
1
of
1
results
1
Promotions Available
3-Methoxybenzenesulfonamide, 97%, Thermo Scientific Chemicals
3-Methoxybenzenesulfonamide, 97%, Thermo Scientific Chemicals
CAS: 58734-57-9 Molecular Formula: C7H9NO3S Molecular Weight (g/mol): 187.21 MDL Number: MFCD08704579 InChI Key: VBKIEQKVSHDVGH-UHFFFAOYSA-N PubChem CID: 14763060 IUPAC Name: 3-methoxybenzenesulfonamide SMILES: COC1=CC(=CC=C1)S(N)(=O)=O