CAS RN 5874-75-9
IUPAC Name:
methyl 2-amino-5-{[(benzyloxy)carbonyl]amino}pentanoate hydrochloride
Synonyms:
methyl 2-amino-5-{[(benzyloxy)carbonyl]amino}pentanoate hydrochloride
Molecular Weight (g/mol):
316.78
Molecular Formula:
C14H21ClN2O4
InChi Key:
ZUJMKNWYEVTCNT-UHFFFAOYNA-N
SMILES:
Cl.COC(=O)C(N)CCCNC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
Ndelta-Benzyloxycarbonyl-L-ornithine methyl ester hydrochloride, 98%, Thermo Scientific™
CAS: 5874-75-9 Molecular Formula: C14H21ClN2O4 Molecular Weight (g/mol): 316.78 MDL Number: MFCD00077027 InChI Key: ZUJMKNWYEVTCNT-UHFFFAOYNA-N Synonym: h-orn z-ome.hcl,h-orn z-ome hcl,h-orn z-ome-hcl,h-orn z-ome hydrochloride,c14h20n2o4.hcl,n-delta-z-l-ornithine methyl ester hydrochloride,h-orn cbz-ome inverted exclamation mark currencyhcl,methyl 2s-2-amino-5-benzyloxycarbonylaminopentanoate hydrochloride,l-ornithine,n5-phenylmethoxy carbonyl-, methyl ester, hydrochloride 1:1,methyl 2s-2-amino-5-benzyloxy carbonyl amino pentanoate hydrochloride PubChem CID: 13967650 SMILES: Cl.COC(=O)C(N)CCCNC(=O)OCC1=CC=CC=C1