CAS RN 58749-22-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Licochalcone-A, Synthetic
An estrogenic flavonoid with anti-tumor and antiparasitic properties.