CAS RN 58753-54-1
IUPAC Name:
hydrogen 3-[benzyl(propan-2-yl)amino]-1-(naphthalen-2-yl)propan-1-one chloride
Synonyms:
hydrogen 3-[benzyl(isopropyl)amino]-1-(naphthalen-2-yl)propan-1-one chloride
Molecular Weight (g/mol):
367.92
Molecular Formula:
C23H26ClNO
InChi Key:
NJTUORMLOPXPBY-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC(C)N(CCC(=O)C1=CC=C2C=CC=CC2=C1)CC1=CC=CC=C1
1
–
1
of
1
results
1
ZM 39923 hydrochloride, R&D Systems™
CAS: 58753-54-1 Molecular Formula: C23H26ClNO Molecular Weight (g/mol): 367.917 InChI Key: NJTUORMLOPXPBY-UHFFFAOYSA-N PubChem CID: 176406 IUPAC Name: 3-[benzyl(propan-2-yl)amino]-1-naphthalen-2-ylpropan-1-one;hydrochloride SMILES: CC(C)N(CCC(=O)C1=CC2=CC=CC=C2C=C1)CC3=CC=CC=C3.Cl