accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58755-70-7

IUPAC Name: 1-iodo-4-methoxy-2-nitrobenzene
Synonyms: 1-iodo-4-methoxy-2-nitrobenzene
Molecular Weight (g/mol): 279.03
Molecular Formula: C7H6INO3
InChi Key: JWZODIRSJJQOKY-UHFFFAOYSA-N
SMILES: COC1=CC=C(I)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results