accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBrOCl

CAS RN 58777-57-4

IUPAC Name: 3-bromofuran-2-carbonyl chloride
Synonyms: 3-bromofuran-2-carbonyl chloride
Molecular Weight (g/mol): 209.42
Molecular Formula: C5H2BrClO2
InChi Key: WTZDPJVRZQGTET-UHFFFAOYSA-N
SMILES: ClC(=O)C1=C(Br)C=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results