accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHN

CAS RN 588-46-5

IUPAC Name: N-benzylacetamide
Synonyms: N-benzylacetamide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: UZJLYRRDVFWSGA-UHFFFAOYSA-N
SMILES: CC(=O)NCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)