accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrO

CAS RN 588-63-6

IUPAC Name: (3-bromopropoxy)benzene
Synonyms: benzene, (3-bromopropoxy)-
Molecular Weight (g/mol): 215.09
Molecular Formula: C9H11BrO
InChi Key: NIDWUZTTXGJFNN-UHFFFAOYSA-N
SMILES: BrCCCOC1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)