CAS RN 588-64-7
IUPAC Name:
(2E)-1-phenyl-2-(phenylmethylidene)hydrazine
Synonyms:
(2E)-1-phenyl-2-(phenylmethylidene)hydrazine
Molecular Weight (g/mol):
196.25
Molecular Formula:
C13H12N2
InChi Key:
JGOAZQAXRONCCI-SDNWHVSQSA-N
SMILES:
N(\N=C\C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Benzaldehyde Phenylhydrazone 98.0+%, TCI America™
CAS: 588-64-7 Molecular Formula: C13H12N2 Molecular Weight (g/mol): 196.253 MDL Number: MFCD00051318 InChI Key: JGOAZQAXRONCCI-SDNWHVSQSA-N PubChem CID: 6436752 IUPAC Name: N-[(E)-benzylideneamino]aniline SMILES: C1=CC=C(C=C1)C=NNC2=CC=CC=C2