accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOFFF

CAS RN 58808-59-6

IUPAC Name: 4-(2,2,2-trifluoroacetyl)benzoic acid
Synonyms: 4-(2,2,2-trifluoroacetyl)benzoic acid
Molecular Weight (g/mol): 218.13
Molecular Formula: C9H5F3O3
InChi Key: WLTZCRCZDLLXQP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results