accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 58823-95-3

IUPAC Name: barium(2+) [(2R,3S,4S,5R,6R)-3,4,5,6-tetrahydroxyoxan-2-yl]methyl phosphate
Synonyms: barium(2+) β-D-glucose 6-phosphate(2-)
Molecular Weight (g/mol): 395.45
Molecular Formula: C6H11BaO9P
InChi Key: FFNFQIZRCUHVAP-WYRLRVFGSA-L
SMILES: [Ba++].O[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results