CAS RN 58861-48-6
IUPAC Name:
2-(4-fluorophenyl)thiophene
Synonyms:
2-(4-fluorophenyl)thiophene
Molecular Weight (g/mol):
178.22
Molecular Formula:
C10H7FS
InChi Key:
PURJRGMZIKXDMW-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1)C1=CC=CS1
1
–
1
of
1
results
1
2-(4-Fluorophenyl)thiophene 95.0+%, TCI America™
CAS: 58861-48-6 Molecular Formula: C10H7FS Molecular Weight (g/mol): 178.224 MDL Number: MFCD06802535 InChI Key: PURJRGMZIKXDMW-UHFFFAOYSA-N PubChem CID: 21894783 IUPAC Name: 2-(4-fluorophenyl)thiophene SMILES: C1=CSC(=C1)C2=CC=C(C=C2)F