accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOONSOOFFFFFFK

CAS RN 588668-97-7

IUPAC Name: potassium 4,4,5,5,6,6-hexafluoro-1,1,3,3-tetraoxo-1λ⁶,3λ⁶,2-dithiazinan-2-ide
Synonyms: potassium 4,4,5,5,6,6-hexafluoro-1,1,3,3-tetraoxo-1λ⁶,3λ⁶,2-dithiazinan-2-ide
Molecular Weight (g/mol): 331.24
Molecular Formula: C3F6KNO4S2
InChi Key: XHXQVKHBZXYEKL-UHFFFAOYSA-N
SMILES: [K+].FC1(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C1(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results