accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOHNOH

CAS RN 58885-58-8

IUPAC Name: tert-butyl N-(3-hydroxypropyl)carbamate
Synonyms: tert-butyl N-(3-hydroxypropyl)carbamate
Molecular Weight (g/mol): 175.23
Molecular Formula: C8H17NO3
InChi Key: XDJCYKMWJCYQJM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCO
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)