accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOF

CAS RN 589-06-0

IUPAC Name: 4-(4-fluorophenyl)butanoic acid
Synonyms: 4-(4-fluorophenyl)butanoic acid
Molecular Weight (g/mol): 182.19
Molecular Formula: C10H11FO2
InChi Key: XVQYBBYOYJXQBF-UHFFFAOYSA-N
SMILES: OC(=O)CCCC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results