accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CNH

CAS RN 589-09-3

IUPAC Name: N-(prop-2-en-1-yl)aniline
Synonyms: N-allyl-aniline
Molecular Weight (g/mol): 133.19
Molecular Formula: C9H11N
InChi Key: LQFLWKPCQITJIH-UHFFFAOYSA-N
SMILES: C=CCNC1=CC=CC=C1
Boiling Point 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (6)
  • (9)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)