accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 589-63-9

IUPAC Name: octan-4-one
Synonyms: 4-octanone
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: YWXLSHOWXZUMSR-UHFFFAOYSA-N
SMILES: CCCCC(=O)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results