accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3N

CAS RN 589-93-5

IUPAC Name: 2,5-dimethylpyridine
Synonyms: 2,5-dimethylpyridine
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: XWKFPIODWVPXLX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)N=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)