accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNHOOHNS

CAS RN 58909-39-0

IUPAC Name: 2-methyl-3-sulfanylidene-1,2,4-triazinane-5,6-dione
Synonyms: 2-methyl-3-sulfanylidene-1,2,4-triazinane-5,6-dione
Molecular Weight (g/mol): 159.16
Molecular Formula: C4H5N3O2S
InChi Key: UMWWHOXOVPIGFD-UHFFFAOYSA-N
SMILES: CN1NC(=O)C(=O)NC1=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results