accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHNOHNOCH3

CAS RN 5893-10-7

IUPAC Name: N-(3-ethoxypropyl)-5-(4-hexylbenzamido)-2-(pyrrolidin-1-yl)benzamide
Synonyms: N-(3-ethoxypropyl)-5-(4-hexylbenzamido)-2-(pyrrolidin-1-yl)benzamide
Molecular Weight (g/mol): 479.67
Molecular Formula: C29H41N3O3
InChi Key: NYGKTOCEZBEXDC-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C(=O)NC1=CC=C(N2CCCC2)C(=C1)C(=O)NCCCOCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results