accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5896-17-3

IUPAC Name: 2-(benzyloxy)benzaldehyde
Synonyms: 2-(benzyloxy)benzaldehyde
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: PBEJTRAJWCNHRS-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)